Labx™170 Cleanroom Wipers
Labx™170 Cleanroom Wipers
Actual product may vary
Labx™170 Cleanroom Wipers
For ISO Class 6 (Class 1000) applications
Have Questions?
Catalog NumberDimensions (L x W)
BERLB1700909409 x 9 in. (22.86 x 22.86 cm)
Catalog number BERLB170090940
Price (SGD)
-
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
  • Made from 80% abaca and 20% wood pulp
  • Cleaner alternative to industrial tissues and wipers
  • Excellent for use as a wafer separator, wet bench liner, laboratory workmat, material packaging and blotter
  • Offer great sorptive capacity
  • Contain no surfactants and virtually no organic extractables
  • 100% biodegradeable
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)22.86 cm
Width (English)9 in.
Width (Metric)22.86 cm
MaterialWood Pulp
AbsorbancyHighly absorbent
Basis Weight17.6
Certifications/ComplianceRoHS 2002/05, REACH
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)For ISO Class 6 cleanroom applications
For Use With (Equipment)Lightweight wafer separator material, wiping of catheters, heart valves and other biomedical devices, cleaning of lab equipment
Unit SizeCS