Owl™ Leveling Platform
Owl™ Leveling Platform
Actual product may vary
Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Have Questions?
Catalog NumberType
BBL-1Bubble Level
Catalog number BBL-1
Price (SGD)
41.00
EA
Add to cart
Type:
Bubble Level
Price (SGD)
41.00
EA
Add to cart
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
  • Size: 36 x 46cm
  • Includes bubble level
Specifications
For Use With (Equipment)Owl System
Product LineOwl
TypeBubble Level
Unit SizeEA