Transfer Vessels Withdrawal Device
Transfer Vessels Withdrawal Device
Actual product may vary
Transfer Vessels Withdrawal Device
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
AY509X5LN2 Transfer Vessels, For Thermo 10
AY509X3LN2 Transfer Vessels, For Thermo 30
AY509X4LN2 Transfer Vessels, For Thermo 20
Catalog number AY509X5
Price (SGD)
2,083.00
EA
Add to cart
For Use With (Equipment):
LN2 Transfer Vessels, For Thermo 10
Price (SGD)
2,083.00
EA
Add to cart
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
  • Available for 10, 20 and 30L models
  • Warranty varies by country, please contact us for details
  • Specifications
    TypeLN2 Withdrawal Device, 8L/min.
    Capacity (English)1.05 cu. ft.
    Capacity (Metric)30 L
    For Use With (Equipment)LN2 Transfer Vessels, For Thermo 10
    Unit SizeEA