anti-CD11c, Clone: 3.9
Product Image
anti-CD11c, Clone: 3.9
Have Questions?
Catalog number ASO3016-08
Price (SGD)
-
Host Species: Mouse
Clone: 3.9
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
AntigenCD11c
ClassificationMonoclonal
Clone3.9
ConjugatePE-Cyanine7
Host SpeciesMouse
Purification MethodUnpurified
Quantity100 Tests
Primary or SecondaryPrimary
Target SpeciesHuman
IsotypeIgG1 κ
Unit SizeEach