Check Order Status
Class B Glass Burettes
Actual product may vary
Class B Glass Burettes
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Rogo Sampaic™ Class B Glass Burettes are manufactured from durable clear borosilicate glass.
Have Questions?
Catalog Number
Capacity (Metric)
ASO1550-03
50 mL
Catalog number
ASO1550-03
Price (SGD)
-
Capacity (Metric):
50 mL
With funnel
Straight glass faucet
Permanent red graduation marks
Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric)
50 mL
Material
Borosilicate Glass
Graduations
0.10 mL
Tolerance
0.10 mL