MICROFLEX™ 93-260 Polychloroprene Gloves
MICROFLEX™ 93-260 Polychloroprene Gloves
Actual product may vary
MICROFLEX™ 93-260 Polychloroprene Gloves
MICROFLEX™ 93-260 Polychloroprene Gloves
MICROFLEX™ 93-260 Polychloroprene Gloves
Polychloroprene disposable gloves offer tactility, dexterity, comfort, and tough chemical resistance. Ideal for aerospace, automotive, life science, and chemical industries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
ANL93260090Large
ANL93260070Small
ANL93260080Medium
Catalog number ANL93260090
Price (SGD)
-
Size:
Large
Polychloroprene disposable gloves offer tactility, dexterity, comfort, and tough chemical resistance. Ideal for aerospace, automotive, life science, and chemical industries.
  • Three-layer design: Offering resistance to harsh chemicals
  • Thinnest chemical-resistant disposable glove: For tactility and dexterity
  • Low AQL: 0.65 AQL for allowable pinholes means exceptional barrier integrity
N/A
Specifications
Certifications/ComplianceFDA 21 CFR 177.2600, ASTM D6978-05, ASTM F1671
Certifications/Compliance (Europe)Category III, EAC TP TC 019:2011, EN 420:2003 + A1:2009
ChlorinatedNo
ApplicationsLaboratory
AQL0.65
ColorGreen
Finger Thickness (English)7.9 mil
Finger Thickness (Metric)0.20 mm
Latex FreeYes
MaterialNeoprene, Nitrile
Palm Thickness (English)7.9 mil
Palm Thickness (Metric)0.198 mm
SizeLarge
Thickness (English)7.9 mil
Thickness (Metric)0.20 mm
EN388 StandardEN 388:2003, EN 388:2016
EN 421Yes
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESJKLOPST
EN ISO 374 - 5Bacteria, Fungi WITH Virus Protection
Glove Cuff StyleBeaded
LiningUnlined
Manufacturing/QMS Audit StandardsISO 9001
Model93-260
SeriesMICROFLEX™ 93-260
Length (English)11.8 in.
Length (Metric)30 cm
Unit SizeCS