▸
Personal Protective Equipment
▸
Gloves
▸
Chemical Resistant Gloves
▸
TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
Actual product may vary
TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
Have Questions?
Change view
Catalog Number
Size
ANL92-605-M-BX
8.5/9
ANL92-605-S-BX
6.5/7
FIS19-148-540
6.5/7
ANL92-605-L-BX
7.5/8
FIS19-148-541
7.5/8
FIS19-148-542
8.5/9
ANL92-605-XL-BX
9.5/10
7 Options
Catalog number
ANL92-605-M-BX
Price (SGD)
-
Size:
8.5/9
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
Does not contain natural rubber latex proteins
Strong grip in wet or dry conditions
Rolled beaded for easy donning
Powder free, Polymer-coated
Ambidextrous
Textured Fingers
AQL: 1.5
Specifications
Chlorinated
Yes
AQL
1.5
Color
Green
Disposable
Single-use
Finger Thickness (English)
6.2 mil
Finger Thickness (Metric)
0.16 mm
Latex Free
Yes
Palm Thickness (English)
4.7 mil
Palm Thickness (Metric)
0.12 mm
Size
8.5/9
EN 421
Yes
EN ISO 374-1
Type B
Glove Cuff Style
Beaded/Rolled
Unit Size
Box