TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves
Actual product may vary
TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves
Sterile neoprene (polychloroprene) cleanroom gloves are free of proteins and accelerators. Suitable for use in aseptic and Class 100/ISO 5 cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
ANL737010606
FIS19-130-08756
ANL737010656.5
FIS19-130-08766.5
ANL737010707
FIS19-130-08777
ANL737010757.5
FIS19-130-08787.5
FIS19-130-08798
ANL737010858.5
FIS19-130-08808.5
FIS19-130-08819
Catalog number ANL73701060
Price (SGD)
460.00
Special offer
Ends: 31-Dec-2026
555.00
Save 95.00 (17%)
Case
Size:
6
Price (SGD)
460.00
Special offer
Ends: 31-Dec-2026
555.00
Save 95.00 (17%)
Case
  • Sterile disposable gloves feature a unique neoprene formulation for greater durability and puncture resistance.
  • Offers high levels of chemical splash resistance protecting from various hazards.
  • Beaded cuff designed with SUREFIT™ technology prevents cuffs from rolling down.
  • Latex-free and chemical accelerator-free eliminates Type I and Type IV allergy risks.
  • Recommended for transferring liquids and solids, sample taking, blending, compounding, and aseptic filling/labeling.
Specifications
Certifications/Compliance21 CFR 820 Quality System Regulations, ASTM D3577, ASTM D7160, ASTM D6978-05, ISO 10282, ISO 11193, ISO 13485, ISO 14001
Certifications/Compliance (Europe)CE 0493, EN 420:2003 + A1:2009, EN 421:2010, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016
Chemotherapeutic RatedYes
ChlorinatedYes
Cleanroom ClassClass 100 (ISO 5)
Accelerator FreeYes
AQL0.65
ColorGreen
DisposableSingle-use
Exterior FinishNo Texture
Finger Thickness (English)8.3 mil
Finger Thickness (Metric)0.19 mm
HandSpecific
Latex FreeYes
MaterialPolychloroprene
Palm Thickness (English)7 mil
Palm Thickness (Metric)0.18 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size6
SterilitySterile
Thickness (English)8.3 mil
Thickness (Metric)0.19 mm
Grip DesignTextured Fingers
Manufacturing/QMS Audit StandardsISO 9001:2015
Model73-701
Packaging TypeCase
SeriesTouchNTuff 73-701
Tensile Strength17 MPa
Length (English)11.6 in.
Length (Metric)29.5 cm
Unit SizeCase