Hydrogen peroxide, 12% w/v
Hydrogen peroxide, 12% w/v
Thermo Scientific Chemicals
Hydrogen peroxide, 12% w/v
CAS: 7722-84-1 | H2O2 | 34.01 g/mol
Have Questions?
Catalog NumberQuantity
ACO4645700101 L
Catalog number ACO464570010
Price (SGD)
66.40
Special offer
Ends: 31-Mar-2026
83.00
Save 16.60 (20%)
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
66.40
Special offer
Ends: 31-Mar-2026
83.00
Save 16.60 (20%)
EA
Add to cart
Specifications
Chemical Name or MaterialHydrogen peroxide
CAS7722-84-1
Boiling Point100.0°C
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement May intensify fire; oxidizer.