Sodium azide, 99+%, for biochemistry
Sodium azide, 99+%, for biochemistry
Sodium azide, 99+%, for biochemistry
Sodium azide, 99+%, for biochemistry
Thermo Scientific Chemicals
Sodium azide, 99+%, for biochemistry
Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain. | CAS: 26628-22-8 | N3Na | 65.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACO447815000
Price (SGD)
376.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
376.00
EA
Add to cart
Specifications
Boiling Point300.0°C
Chemical Name or MaterialSodium azide
Melting Point275.0°C
CAS26628-22-8
Health Hazard 1GHS Signal Word: Danger
View more
General Information
• Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
• One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation
Applications
• Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
• Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
• Sodium azide is used for the introduction of an azide group to prepare a desired compound
• It is useful in biochemical approaches as a probe reagent and a preservative
• This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates
RUO – Research Use Only
General References:
  1. Abdollahi-Alibeik, M.; Moaddeli, A. Multi-component one-pot reaction of aldehyde, hydroxylamine and sodium azide catalyzed by Cu-MCM-41 nanoparticles: a novel method for the synthesis of 5-substituted 1 H-tetrazole derivatives. New J. Chem. 2015, 39 (3), 2116-2122.
  2. Zeng, M.; Yang, Y. H.; Li, J. J.; Chen, Y.; Cui, D. M.; Zhang, C. Copper Catalyzed Synthesis of Aryl Azides from Aryl Bromides and Sodium Azide. Asian J. Chem. 2015, 27 (5), 1698-1700.