Potassium phosphate, dibasic, trihydrate, extra pure
CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACO447732500
250 g
Plastic bottle
Catalog number ACO447732500
Price (SGD)
70.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
70.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium phosphate, dibasic, trihydrate
CAS16788-57-1
Quantity250 g
Molecular FormulaH7K2O7P
Linear FormulaK2HPO4·3H2O