Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Thermo Scientific Chemicals
Citric acid, trisodium salt dihydrate, ACS reagent
CAS: 6132-04-3 | C6H5Na3O7·2H2O | 294.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO446332500250 gPlastic Bottle
Catalog number ACO446332500
Price (SGD)
56.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
56.00
EA
Add to cart
Specifications
Chemical Name or MaterialCitric Acid, Trisodium Salt Dihydrate
CAS6132-04-3
Melting Point>300.0°C
PackagingPlastic Bottle
Molecular FormulaC6H5Na3O7·2H2O