Potassium hydrogen carbonate, ACS reagent
Potassium hydrogen carbonate, ACS reagent
Potassium hydrogen carbonate, ACS reagent
Potassium hydrogen carbonate, ACS reagent
Thermo Scientific Chemicals
Potassium hydrogen carbonate, ACS reagent
CAS: 298-14-6 | CHKO3 | 100.11 g/mol
Have Questions?
Catalog NumberQuantity
ACO446305000500 g
Catalog number ACO446305000
Price (SGD)
71.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
71.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen carbonate
Melting Point292.0°C
CAS298-14-6
Quantity500 g
EINECS Number206-059-