Ethanol, 70%, technical, denaturated with 5% methanol
Ethanol, 70%, technical, denaturated with 5% methanol
Ethanol, 70%, technical, denaturated with 5% methanol
Ethanol, 70%, technical, denaturated with 5% methanol
Thermo Scientific Chemicals
Ethanol, 70%, technical, denaturated with 5% methanol
CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4442000505 LPlastic Drum
Catalog number ACO444200050
Price (SGD)
177.60
Special offer
Ends: 31-Mar-2026
222.00
Save 44.40 (20%)
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (SGD)
177.60
Special offer
Ends: 31-Mar-2026
222.00
Save 44.40 (20%)
EA
Add to cart
Specifications
Chemical Name or MaterialEthanol
Melting Point-114°C
CAS64-17-5
Boiling Point78°C
Quantity5 L
View more
  • Ethanol, 70%, technical, denaturated 5L with 5% methanol Plastic Bottle Cas No. 64-17-5
For Manufacturing and Laboratory Use. Not for beverage use