4-Fluorobenzaldehyde, 98+%, AcroSeal™
4-Fluorobenzaldehyde, 98+%, AcroSeal™
4-Fluorobenzaldehyde, 98+%, AcroSeal™
4-Fluorobenzaldehyde, 98+%, AcroSeal™
Thermo Scientific Chemicals
4-Fluorobenzaldehyde, 98+%, AcroSeal™
CAS: 459-57-4 | C7H5FO | 124.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO427191000100 mLAcroSeal™ Glass bottle
Catalog number ACO427191000
Price (SGD)
248.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (SGD)
248.00
Each
Specifications
Chemical Name or Material4-Fluorobenzaldehyde
CAS459-57-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0