Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO424575000500 gPlastic bottle
Catalog number ACO424575000
Price (SGD)
180.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
180.00
EA
Add to cart
Specifications
Absorbance0.2 max. at 290nm, (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm
Chemical Name or MaterialTris(hydroxymethyl)aminomethane
Absorbance Conditions(40% aq. soln.) (1 cm cell)
CAS77-86-1
Melting Point>168.5°C
View more
Biological buffer
RUO – Research Use Only