Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Thermo Scientific Chemicals
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4245500101 LEcoSafPak™ Glass Bottle
Catalog number ACO424550010
Price (SGD)
92.00
EA
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (SGD)
92.00
EA
Add to cart
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Density(20/20°C) 0.865 to 0.870
Boiling point109°C to 111°C
Residue after evaporation=<0.001 %
Sulfur compounds (as S)=<0.003 %