Thioacetamide, 99+%, ACS reagent
Thioacetamide, 99+%, ACS reagent
Thioacetamide, 99+%, ACS reagent
Thioacetamide, 99+%, ACS reagent
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO42453025025 gGlass bottle
Catalog number ACO424530250
Price (SGD)
137.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
137.00
EA
Add to cart
Specifications
Chemical Name or MaterialThioacetamide
CAS62-55-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water