Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Thermo Scientific Chemicals
Sulfuric acid, ACS reagent, 95% solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO424525000500 mLGlass Bottle
Catalog number ACO424525000
Price (SGD)
97.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
97.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Melting Point10.0°C
Boiling Point290.0°C
Health Hazard 1Danger