Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change view
Quantity:
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACO424375000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate, dibasic
CAS7558-79-4
ColorWhite
Health Hazard 1Signal Word: CAUTION!
pH8.7 to 9.3 (5% soln. at 25°C)