Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Thermo Scientific Chemicals
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO424061000100 gPlastic bottle
Catalog number ACO424061000
Price (SGD)
56.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
56.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen phthalate
CAS877-24-7
Melting Point295.0°C to 300.0°C
ColorWhite
RTECS NumberCZ4326000