N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, C12H14N2.2ClH, CAS Number-1465-25-4 | CAS: 1465-25-4 | C12H16Cl2N2 | 259.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO42399025025 gGlass Bottle
Catalog number ACO423990250
Price (SGD)
275.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
275.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-(1-Naphthyl)ethylenediamine, dihydrochloride
CAS1465-25-4
CAS Min %98.0
Health Hazard 1Warning
CAS Max %100.0