Hydrochloric acid, ACS reagent, ca. 37% solution in water
Hydrochloric acid, ACS reagent, ca. 37% solution in water
Hydrochloric acid, ACS reagent, ca. 37% solution in water
Hydrochloric acid, ACS reagent, ca. 37% solution in water
Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number ACO423795000
Price (SGD)
67.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
67.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric acid
Name Noteca. 37% Solution in Water
CAS7647-01-0
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Density1.1800g/mL