Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4237300505 gGlass Bottle
Catalog number ACO423730050
Price (SGD)
56.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
56.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron(II) sulfate heptahydrate
Melting Point64.0°C
CAS7782-63-0
ColorBlue or Blue-Green or Green
Health Hazard 1GHS Signal Word: Warning