Iron(III) chloride hexahydrate, 97+%, ACS reagent
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Thermo Scientific Chemicals
Iron(III) chloride hexahydrate, 97+%, ACS reagent
CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO423705000500 gPlastic Bottle
Catalog number ACO423705000
Price (SGD)
154.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
154.00
EA
Add to cart
Specifications
Boiling Point280.0°C to 285.0°C
Chemical Name or MaterialIron(III) chloride hexahydrate
Melting Point37.0°C
CAS10025-77-1
ColorWhite
View more
Lewis acid
RUO – Research Use Only