Diphenylamine, ACS reagent
Diphenylamine, ACS reagent
Diphenylamine, ACS reagent
Diphenylamine, ACS reagent
Thermo Scientific Chemicals
Diphenylamine, ACS reagent
CAS: 122-39-4 | C12H11N | 169.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO423651000100 gGlass bottle
Catalog number ACO423651000
Price (SGD)
132.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
132.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiphenylamine
Name NoteFor analysis ACS
CAS122-39-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
View more
Indicator
RUO – Research Use Only