Phthalic anhydride, ACS reagent
Phthalic anhydride, ACS reagent
Phthalic anhydride, ACS reagent
Phthalic anhydride, ACS reagent
Thermo Scientific Chemicals
Phthalic anhydride, ACS reagent
CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO423325000500 gGlass bottle
Catalog number ACO423325000
Price (SGD)
117.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
117.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhthalic anhydride
CAS85-44-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat