Ammonium molybdate(VI) tetrahydrate, ACS reagent
Ammonium molybdate(VI) tetrahydrate, ACS reagent
Ammonium molybdate(VI) tetrahydrate, ACS reagent
Ammonium molybdate(VI) tetrahydrate, ACS reagent
Thermo Scientific Chemicals
Ammonium molybdate(VI) tetrahydrate, ACS reagent
CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO423311000100 gPlastic bottle
Catalog number ACO423311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium molybdate(VI) tetrahydrate
Melting Point190°C
CAS12054-85-2
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
Oxidation reagent
RUO – Research Use Only