Ammonium hydroxide, ACS reagent, 28-30% solution in water
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Thermo Scientific Chemicals
Ammonium hydroxide, ACS reagent, 28-30% solution in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ACO4233000252.5 L
Catalog number ACO423300025
Price (SGD)
189.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
189.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
ColorColorless
Boiling Point38.0°C
CAS1336-21-6
Health Hazard 1Danger