Sodium metabisulfite, 97+%, ACS reagent
Sodium metabisulfite, 97+%, ACS reagent
Sodium metabisulfite, 97+%, ACS reagent
Sodium metabisulfite, 97+%, ACS reagent
Thermo Scientific Chemicals
Sodium metabisulfite, 97+%, ACS reagent
CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO419582500250 gPlastic bottle
Catalog number ACO419582500
Price (SGD)
75.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
75.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium metabisulfite
Melting Point150.0°C
CAS7681-57-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Reduction reagent
RUO – Research Use Only