Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Thermo Scientific Chemicals
Sodium hypochlorite, 4-6%
CAS: 7681-52-9 | ClNaO | 74.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4195500101 LPlastic bottle
Catalog number ACO419550010
Price (SGD)
124.00
EA
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
124.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium hypochlorite
Name Note4-6% Chlorine
CAS7681-52-9
ColorYellow
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only