Mineral oil, pure
Thermo Scientific Chemicals
Mineral oil, pure
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACO415080025
Price (SGD)
160.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
160.00
EA
Add to cart
Specifications
Chemical Name or MaterialMineral Oil
CAS8042-47-5
ColorColorless
Density0.8770g/mL
EINECS Number232-455-8