Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Thermo Scientific Chemicals
Malachite Green oxalate, pure, certified
CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO413491000100 gGlass bottle
Catalog number ACO413491000
Price (SGD)
235.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
235.00
EA
Add to cart
Specifications
Chemical Name or MaterialMalachite Green oxalate
Name NotePure
CAS2437-29-8
Health Hazard 1Danger
Absorption Ratio1.00 to 1.15