Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Thermo Scientific Chemicals
Isopropyl methanesulfonate, 99%
CAS: 926-06-7 | C4H10O3S | 138.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO41284025025 gGlass bottle
Catalog number ACO412840250
Price (SGD)
498.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
498.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropyl methanesulfonate
Name Note99%
CAS926-06-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.