N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
CAS: 62778-12-5 | 0·5 C2H2O4 | 362.42 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO40849025025 gGlass bottle
Catalog number ACO408490250
Price (SGD)
305.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
305.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethyl-p-phenylenediamine oxalate
Name Note99%
CAS62778-12-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger