Bromocresol Green, ACS reagent
Bromocresol Green, ACS reagent
Bromocresol Green, ACS reagent
Thermo Scientific Chemicals
Bromocresol Green, ACS reagent
CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO40293010010 gGlass bottle
Catalog number ACO402930100
Price (SGD)
139.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
139.00
Each
Specifications
Chemical Name or MaterialBromocresol Green
CAS76-60-8
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Indicator
RUO – Research Use Only