Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Thermo Scientific Chemicals
Ammonium persulfate, 98+%, ACS reagent
Oxidation reagent | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO401165000500 gPlastic bottle
Catalog number ACO401165000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium persulfate
Melting Point120.0°C
CAS7727-54-0
ColorWhite
Health Hazard 1Danger
View more
Suitable for battery materials development.
Oxidation reagent
RUO – Research Use Only