Silicone Grease, medium viscosity, Tube with 35gr
Silicone Grease, medium viscosity, Tube with 35gr
Thermo Scientific Chemicals
Silicone Grease, medium viscosity, Tube with 35gr
Have Questions?
Catalog NumberQuantity
ACO3861100101 Ea.
Catalog number ACO386110010
Price (SGD)
47.20
Special offer
Ends: 31-Mar-2026
59.00
Save 11.80 (20%)
EA
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (SGD)
47.20
Special offer
Ends: 31-Mar-2026
59.00
Save 11.80 (20%)
EA
Add to cart
Specifications
Product TypeSilicone Grease
Quantity1 Ea.
Volume (English)1.23 oz.