Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog NumberQuantity
ACO3839100252.5 L
Catalog number ACO383910025
Price (SGD)
358.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
358.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
CAS67-63-0
Density0.7850g/mL
Boiling Point81.0°C to 83.0°C