Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Thermo Scientific Chemicals
Sodium hydroxide, for analysis, 50 wt% solution in water
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
Catalog number ACO380215000
Price (SGD)
82.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
82.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
ColorColorless
Boiling Point145.0°C
CAS1310-73-2
Health Hazard 1Danger