Triethyl citrate, 99%
Triethyl citrate, 99%
Triethyl citrate, 99%
Triethyl citrate, 99%
Thermo Scientific Chemicals
Triethyl citrate, 99%
CAS: 77-93-0 | C12H20O7 | 276.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3750600101 kgPlastic bottle
Catalog number ACO375060010
Price (SGD)
179.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
179.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriethyl citrate
CAS77-93-0
Melting Point-46.0°C
Density1.1360g/mL
EINECS Number201-070-7