Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Thermo Scientific Chemicals
Sand, pure, 40-100 mesh
CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3709400101 kgPlastic bottle
Catalog number ACO370940010
Price (SGD)
108.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
108.00
EA
Add to cart
Specifications
Boiling Point2230.0°C
Chemical Name or MaterialSand
Melting Point1610.0°C
Name Note40 to 100 Mesh
CAS14808-60-7
View more
Filter aid
RUO – Research Use Only