Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO364395000500 mLGlass Bottle
Catalog number ACO364395000
Price (SGD)
80.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
80.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
Name Notewater <50 ppm
CAS67-56-1