Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number ACO363830100
Price (SGD)
821.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
821.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris(2-carboxyethyl)phosphine hydrochloride
CAS51805-45-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.