Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Thermo Scientific Chemicals
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ACO348455000
Price (SGD)
140.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
140.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108.4°C
Name NoteWater <50 ppm, stabilized
CAS109-99-9