1x1 image pixel for data collection
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Thermo Scientific Chemicals
Iron(III) sulfate pentahydrate, 97%
CAS: 142906-29-4 | Fe2O12S3·5H2O | 489.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO345235000500 gPlastic bottle
Catalog number ACO345235000
Price (SGD)
154.00
EA
Estimated availability date 24-Sep-2025
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
154.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron(III) sulfate pentahydrate
CAS142906-29-4
ColorBeige-Yellow to White
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
View more
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link