Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Thermo Scientific Chemicals
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO343811000100 gGlass bottle
Catalog number ACO343811000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate dibasic dihydrate
Melting Point243.0°C to 245.0°C
Name NoteFor HPLC
CAS10028-24-7