Pyridine, 99.5%, Extra Dry, AcroSeal™
Pyridine, 99.5%, Extra Dry, AcroSeal™
Pyridine, 99.5%, Extra Dry, AcroSeal™
Pyridine, 99.5%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL | CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3394200101 LAcroSeal™ Glass Bottle
Catalog number ACO339420010
Price (SGD)
336.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (SGD)
336.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
CAS110-86-1
ColorColorless
Health Hazard 1Danger