Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3273800101 kgGlass Bottle
Catalog number ACO327380010
Price (SGD)
161.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
161.00
EA
Add to cart
Specifications
Chemical Name or MaterialUrea
Name NoteDNAse, RNAse and Protease Free
CAS57-13-6
Absorbance(8M in H2O), (8 M in H2O) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm
Health Hazard 2GHS H Statement: