Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Isopropanol, 99.8%, Extra Dry, AcroSeal™
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog NumberQuantity
ACO3269600101 L
Catalog number ACO326960010
Price (SGD)
287.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
287.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
CAS67-63-0
ColorColorless
Health Hazard 1Danger